Uvex™ Futura™ Goggles
180° panoramic view for better peripheral vision
$33.49 - $33.49
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
S47619C
|
Fisher Science Education
S47619C |
Safety goggles |
Each for $33.49
Pack of 10 Each for $334.90 |
|
Description
- Firm, comfortable seal protects against chemicals, dust, and grindings
- Impact-resistant, chemical-splash polycarbonate lens
- Permanently bonded uvextreme™ AF antifog coating
- Wide, neoprene strap is slotted to prevent slippage
Compliance
Meet ANSI Z87.1-2010 and CSA Z94.3 standards