Honeywell™ Uvex™ Flex Seal™ Goggles
Exceptionally tight seal for protection from impacts and chemical splashes.
Manufacturer: Honeywell™ UVS3405X
Description
- Contemporary
- Low-profile
- With wrap-around
- Optically correct polycarbonate lens configuration increases visibility and protection
- Uvextreme™ antifog coating and indirect ventilation minimize fogging
- Temperature-resistant blue silicone frames fit over virtually all prescription eyewear
- Pivoting clips permit goggle to be positioned on hardhat or on headband above the ears
Order Info
Available with neoprene band or fabric band.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Blue | |
Fabric headband | |
Silicone | |
Clear | |
Polycarbonate |
Fabric | |
Indirect | |
Uvextreme Anti-fog | |
Anti-fog | |
Safety goggles |