Honeywell™ Uvex™ Falcon Replacement Lenses
Sleek, frameless design with wrap-around 9-base unilens. Lightweight comfort and superior coverage providing complete and unobstructed vision.
Manufacturer:  Honeywell™ UVS6700X
 View more versions of this product
Catalog No. 19-055-452
Description
Description
Lighter than air, the Uvex Falcon soars above the competition as a great value in protective eyewear. The Uvex Falcon's frameless and optically perfected dual nine-base wrap-around lens provides superior coverage and unobstructed peripheral vision.
  • Sleek, frameless design with wrap-around 9-base unilens
  • Easy and economical lens replacement system
  • Box of 10 clear replacement lenses with Ultra-dura Anti-scratch Coating
  • Meets ANSI Z87.1-2003 (High Impact) and tested to meet the requirements of the CSA Z94.3 standard
Specifications
Specifications
Replacement lens
Clear
Safety
XTR
Replacement lens
Documents