Uvex™ ExtremePro™ Eyewear
Stands up to the toughest fogging, scratching and sweating
$14.14 - $14.14
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003 and certified to CSA Z94.3 standard
Frame Color Black; Gray
Frame Material Nylon
Includes Moisture-wicking brow
Lens Material Polycarbonate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Lens Coating Lens Tint Lens Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Lens Coating Lens Tint Lens Type Price Quantity  
18-813-467 North Safety Products
SX0301
Minimizes strong outdoor sunlight and glare Supra-Dura Anti-scratch Espresso Scratch-resistant Each for $14.14
Specifications
Description
  • FlexFit wire core temples bend and flex for a customizable, full-comfort fit
  • Flexible and replaceable nosepiece conforms to virtually any nasal profile and provides a contoured, non-slip fit
  • Replaceable MoistureWick™ brow accessory wicks away moisture to keep lenses clear in high humidity and heat
  • GripClick™ frame adjustment lever allows wearer to customize the width of the frame
  • Black/gray frame
Used for foundries, chemical plants, metalworking, machining, pulp and paper, shipbuilding or oil and gas industries
Compliance
Exceeds ANSI™ Z87.1-2003.
Description & Specifications
Specifications
Meets ANSI Z87.1-2003 and certified to CSA Z94.3 standard
Nylon
Polycarbonate
Ultra anti-scratch; Lifetime frame guarantee
Black; Gray
Moisture-wicking brow
Ratcheting