Honeywell Uvex™ Stealth Safety Goggles with Hydroshield™ Anti-Fog Lens Coating
Delivers consistent anti-fog protection that lasts 90x longer
Supplier: Honeywell S3960HSI
Description
- Provides >180 seconds
- Fog free time
- Provides 2x scratch resistance
- >99.9% UVA/UVB protection
- Lifetime frame guarantee
- Lens replacement lenses available
- Indirect ventilation system
- D3 certified
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
HydroShield AF | |
Clear | |
Fabric |
Gray | |
Safety goggles |