Honeywell™ Disposable Face Shields
Designed for rugged jobs—tough yet lightweight and comfortable
Manufacturer: Honeywell™ UVS8565
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
- Shell offers built-in chin protection and extended top-of-head coverage
- Adjustable headgear tilts visor into 2784 possible positions
- Ratcheting headgear features a locking mechanism for secure fit
- Comfortable cell foam on back of headgear with breathable, removable, washable headband
- 100% dielectric; no metal parts
Replacement Visors
- Clear lens suits most indoor work applications
- Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
ANSI Z87.1-2010, CSA Z94.3 | |
Uncoated | |
Replacement visor |
Bionic Face Shield | |
Shade 5 |