Honeywell™ Disposable Face Shields
Designed for rugged jobs—tough yet lightweight and comfortable
Manufacturer:  Honeywell™ UVS8550
 View more versions of this product
Catalog No. 29-635-3814
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
  • Shell offers built-in chin protection and extended top-of-head coverage
  • Adjustable headgear tilts visor into 2784 possible positions
  • Ratcheting headgear features a locking mechanism for secure fit
  • Comfortable cell foam on back of headgear with breathable, removable, washable headband
  • 100% dielectric; no metal parts
Replacement Visors
  • Clear lens suits most indoor work applications
  • Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
Specifications
ANSI Z87.1-2010, CSA Z94.3
Uncoated
Clear
For Bionic facesheild
Polycarbonate
Replacement visor