Honeywell Safety Products™ Uvex™ Astro OTG 3001™ Safety Glasses
Over-the-glasses style
Manufacturer:  Honeywell Safety Products™ UVS2504C
 View more versions of this product
Catalog No. 19-084890
Description
Description
Over-the-glasses style
  • Patented adjustable lens inclination and adjustable temple length offer one-size-fits-all flexibility; Duoflex™ temples have soft comfort cushions for the sensitive mastoid area behind the ear
  • Lightweight, impact-resistant nylon frames feature polycarbonate sideshields and browguard for protection against UV, dust, and flying particles
  • Clear polycarbonate lenses with Ultradura™ hardcoat (UD) that resists scratches and absorbs 99.9% of UV radiation up to 385nm
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Scientific Customer Service Representative at 1-800-772-6733.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Specifications
Specifications
ANSI Z87.1-2015, CSA Z94.3, Military V0 Ballistic Test
Black
Uvextreme Anti-fog
Anti-fog
Gray XTR
Nylon
Gray
OTG safety spectacles