Honeywell™ Uvex™ Ambient OTG Safety Eyewear
Designed to fit over most prescription eyewear, and has a lifetime frame guarantee and economical lens replacement system. Honeywell™ Uvex™ Ambient OTG Safety Eyewear offers exceptional peripheral vision and coverage in a lightweight, wrap-around uni-lens design.
Manufacturer: Honeywell™ S3162
Description
- The Uvex™ Ambient provides over-the-glass coverage, offering your workers full protection while wearing their own prescription eyewear
- Available with a range of comfort features and in two sizes, the Uvex™ Ambient OTG offers all-day coverage for a variety of workers and a wide range of prescription eyewear styles
- This high performance model can improve your workers' productivity as well as lower your cost-of-ownership with Uvex's renowned tints and lens replacement system
- The Uvex™ Ambient™ OTG is lightweight, comfortable and a stylish alternative to prescription safety eyewear
- Exceptional peripheral vision and coverage in a lightweight, wraparound uni-lens design
- Economical and easy lens replacement allows multiple tints and extends the life of the eyewear
Specifications
ANSI Z87.1-2003 and CSA Z94.3 standards | |
Blue | |
Ultra Dura | |
Scratch-resistant |
Minimizes outdoor sunlight and glare | |
Medium | |
Gray | |
Safety Eyewear; Over-the-glasses |