Learn More
Biochrom UV Visible Micro Cell
Accessory for Biochrom WPA units
Supplier:  Biochrom 80210025
Catalog No. BC80-2100-25
Description
Description
  • Micro cell with lid and black walls for visible/UV spectrophotometry.
  • Minimum working volume of 400μL and a 10 mm pathlength.
Specifications
Specifications
UV Visible Micro Cell
Quartz
2 Pk.
Visible/UV Spectrophotometry
400 μL
2 mm
10 mm
Product Suggestions
Product Suggestions