missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ursolic acid, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 290182500
Specifications
Ursolic acid | |
Glass bottle | |
+ 59.00 | |
283.0°C to 285.0°C | |
77-52-1 | |
C30H48O3 | |
15,10075 | |
Solubility in water: insoluble. Other solubilities: soluble in 2% alcoholic naoh,soluble in hot glacial acetic acid,moderately soluble in acetone,slightly soluble in alcohol,ether,chloroform and,carbon disulfide | |
CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1C)C)C(=O)O | |
456.7 | |
456.7 | |
95% |
Authentic | |
+ 59.00 (20.00°C c=0.3,pyridine) | |
White | |
250mg | |
94% min. (HPLC) | |
10,202 | |
3beta-Hydroxyurs-12-en-28-oic acid | |
WCGUUGGRBIKTOS-AABRAELGSA-N | |
(4aS,6aS,6aS,6bR,8aS,12aR,14bR)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid | |
129316832 | |
Crystalline Powder or Needles |
Chemical Identifiers
77-52-1 | |
456.7 | |
3beta-Hydroxyurs-12-en-28-oic acid | |
(4aS,6aS,6aS,6bR,8aS,12aR,14bR)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid |
C30H48O3 | |
WCGUUGGRBIKTOS-AABRAELGSA-N | |
129316832 | |
CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1C)C)C(=O)O |
Safety and Handling
EINECSNumber : 201-034-