missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ursolic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC290180010

 View more versions of this product

Catalog No. AC290180010



Specifications

Specifications

Ursolic acid
Glass bottle
+ 59.00
283.0°C to 285.0°C
77-52-1
C30H48O3
15,10075
Solubility in water: insoluble. Other solubilities: soluble in 2% alcoholic naoh,soluble in hot glacial acetic acid,moderately soluble in acetone,slightly soluble in alcohol,ether,chloroform and,carbon disulfide
CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1C)C)C(=O)O
456.7
456.7
95%
Authentic
+ 59.00 (20.00°C c=0.3,pyridine)
White
1g
94% min. (HPLC)
10,202
3beta-Hydroxyurs-12-en-28-oic acid
WCGUUGGRBIKTOS-AABRAELGSA-N
(4aS,6aS,6aS,6bR,8aS,12aR,14bR)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
129316832
Crystalline Powder or Needles

Chemical Identifiers

77-52-1
456.7
3beta-Hydroxyurs-12-en-28-oic acid
(4aS,6aS,6aS,6bR,8aS,12aR,14bR)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-2,3,4,5,6,6a,7,8,8a,10,11,12,13,14b-tetradecahydro-1H-picene-4a-carboxylic acid
C30H48O3
WCGUUGGRBIKTOS-AABRAELGSA-N
129316832
CC1CCC2(CCC3(C(=CCC4C3(CCC5C4(CCC(C5(C)C)O)C)C)C2C1C)C)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : 201-034-

SDS Documents