Please login to your online account to display your discounted pricing

Alfa Aesar™ Ursodeoxycholic acid, 99%

$45.81 - $45.81

Chemical Identifiers

Chemical Name or Material Ursodeoxycholic acid
CAS 128-13-2
Molecular Formula C24H40O4
Formula Weight 392.56
MDL Number MFCD00003680
Merck Index 14,9889
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049003

  SDS
alfa aesar™
B2049003
1g Each for $45.81
Description & Specifications

Specifications

Formula Weight 392.56
Melting Point 201° to 204°C
Chemical Name or Material Ursodeoxycholic acid
Merck Index 14,9889
Molecular Formula C24H40O4
RTECS Number FZ2000000
Quantity 1g
CAS 128-13-2
Assay 99%
MDL Number MFCD00003680