missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Urotensin II, Goby

For cellular and molecular biology applications

$427.23 - $650.73

Specifications

Product Type Urotensin II, goby
Molecular Weight 1361.57
Storage Requirements -30° to -10°C
Format Solid
Sequence Ala-Gly-Thr-Ala-Asp-Cys-Phe-Trp-Lys-Tyr-Cys-Val, (Disulfide bridge: Cys6-Cys11
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66939LB0 View Documents Alfa Aesar™
J66939LB0
5mg Each for $427.23
 
AAJ66939MA View Documents Alfa Aesar™
J66939MA
10mg Each for $650.73
 
Specifications

Specifications

Urotensin II, goby
-30° to -10°C
Ala-Gly-Thr-Ala-Asp-Cys-Phe-Trp-Lys-Tyr-Cys-Val, (Disulfide bridge: Cys6-Cys11
HPLC
1361.57
Solid
RUO

For Research Use Only