Please login to your online account to display your discounted pricing

Alfa Aesar™ Urocortin III, Mouse, free acid


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66579MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66579MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type Urocortin III, mouse, free acid
Format Solid
Purification Method HPLC
Molecular Weight 4174.00
Storage Requirements -30° to -10°C
Cross Reactivity Mouse
Regulatory Status RUO
Quantity 1mg