missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urocanic acid, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 228960050

 View more versions of this product

Catalog No. AC228960050



Description

Description

Specifications

Specifications

Urocanic acid
0.5% max.
White to Beige
5g
97.5% min. (HPLC)
MFCD00005203
10, 9691
Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
C1=C(NC=N1)C=CC(=O)O
138.13
CHEBI:30818
Powder
Authentic
Glass bottle
217.0°C to 220.0°C
104-98-3
C6H6N2O2
25, 126
2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid, 4-imidazoleacrylic acid, 5-imidazoleacrylic acid, imidazole-4-acrylic acid, imidazoleacrylic acid, trans-urocanic acid, urocanate, urocanic acid, urocaninic acid
LOIYMIARKYCTBW-UPHRSURJSA-N
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
1549103
138.13
98%

Chemical Identifiers

104-98-3
138.13
LOIYMIARKYCTBW-UPHRSURJSA-N
1549103
(Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
C6H6N2O2
MFCD00005203
2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid, 4-imidazoleacrylic acid, 5-imidazoleacrylic acid, imidazole-4-acrylic acid, imidazoleacrylic acid, trans-urocanic acid, urocanate, urocanic acid, urocaninic acid
CHEBI:30818
C1=C(NC=N1)C=CC(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : 203-258-4

SDS Documents