missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Uridine 5'-monophosphate, disodium salt, hydrate, 98%, ACROS Organics™

homogeneous

Manufacturer:  Fisher Scientific Acros 226290010

 View more versions of this product

Catalog No. AC226290010



Description

Description

Specifications

Specifications

0.71 to 0.77 (A250/A260),0.38 to 0.44 (A280/A260)
Authentic
97.5% min. (lambda max. 262nm,pH 7) molar absorptivity 10000+/-2%,on dry basis
26% max. (K.F.)
208.0°C to 210.0°C
3387-36-8
C9H11N2Na2O9P·xH2O
10, 9689
Solubility in water:+/- 40% (20°C)
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)O)O)O.[Na].[Na]
368.14
368.14
98%
Uridine 5′-monophosphate, disodium salt, hydrate
Glass bottle
95% min. (c=1, 430nm) 1cm cell
White
1g
97.5% min. (HPLC)
MFCD00149422
5'-uridylicaciddisodiumsalt, uridine 5'-monophosphate, disodium salt, uridine-5'-monophosphate disodium salt
RSSRHKDOIOBLBS-WFIJOQBCSA-N
[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
123134060
Crystalline Powder

Chemical Identifiers

3387-36-8
368.14
RSSRHKDOIOBLBS-WFIJOQBCSA-N
123134060
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)(O)O)O)O.[Na].[Na]
C9H11N2Na2O9P·xH2O
MFCD00149422
5'-uridylicaciddisodiumsalt, uridine 5'-monophosphate, disodium salt, uridine-5'-monophosphate disodium salt
[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate;sodium
Safety and Handling

Safety and Handling

EINECSNumber : 222-211-9

SDS Documents