missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Uridine-5'-diphosphate trisodium salt, 98+%

$115.81 - $437.96

Chemical Identifiers

CAS 21931-53-3
Molecular Formula C9H11N2Na3O12P2
Molecular Weight (g/mol) 470.106
MDL Number MFCD00006524
InChI Key GFASEGWJDXDCKZ-PRYCXDIZSA-K
Synonym uridine 5'-diphosphate sodium salt, uridine 5'-diphosphate sodium salt from
PubChem CID 126968597
IUPAC Name trisodium;[[(2S,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
SMILES C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])[O-])O)O.[Na+].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60141MC View Documents Alfa Aesar™
J60141MC
100mg Each for $115.81
 
AAJ60141ME View Documents Alfa Aesar™
J60141ME
500mg Each for $437.96
 
Description

Description

Applications
It has been used as a purinergic agonist to study its effects on absorptive cationic flux in cochlear outer sulcus cells (OSC) and vestibular transitional cells (VTC). Also been used for nucleoside diphosphatase (NDPase) activity assays in rabbit retinae.

Solubility
Soluble in water to 100mM

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

21931-53-3
470.106
GFASEGWJDXDCKZ-PRYCXDIZSA-K
126968597
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])[O-])O)O.[Na+].[Na+].[Na+]
C9H11N2Na3O12P2
MFCD00006524
uridine 5'-diphosphate sodium salt, uridine 5'-diphosphate sodium salt from
trisodium;[[(2S,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate

Specifications

White
21931-53-3
MFCD00006524
Soluble in water to 100mM
C1=CN(C(=O)NC1=O)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])[O-])O)O.[Na+].[Na+].[Na+]
470.106
470.11
≥98%
100mg
C9H11N2Na3O12P2
uridine 5'-diphosphate sodium salt, uridine 5'-diphosphate sodium salt from
GFASEGWJDXDCKZ-PRYCXDIZSA-K
trisodium;[[(2S,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate
126968597
Powder
Uridine-5'-diphosphate trisodium salt
Safety and Handling

Safety and Handling

GHS H Statement
H371-H302
May cause damage to organs.
Harmful if swallowed.

GHS P Statement
P260-P264-P309+P311-P301+P312-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C