Learn More
Uridine-5'-diphosphate disodium salt, 98%, Thermo Scientific Chemicals
$142.81 - $197.62
Chemical Identifiers
CAS | 27821-45-0 |
---|---|
Molecular Formula | C9H12N2Na2O12P2 |
Molecular Weight (g/mol) | 448.12 |
MDL Number | MFCD00006524,MFCD00084679,MFCD04975448 |
InChI Key | ZQKVPFKBNNAXCE-HVQNCJTOSA-L |
Synonym | 5'-udp,2na |
PubChem CID | 131676107 |
SMILES | [Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64329MC
|
Thermo Scientific Chemicals
J64329MC |
100 mg |
N/A
|
|
|||||
AAJ6432903
|
Thermo Scientific Chemicals
J6432903 |
1 g |
N/A
|
|
|||||
Description
Uridine-5'-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUridine-5′-diphosphate disodium salt is used in studies on nucleic acid (RNA) biosynthesis and cell signaling. UDP is a nucleotide that upon phosphorylation to UTP becomes a substrate for enzymes such as RNA polymerase(s) and GTPases. These enzymes are involved in a wide range of applications from the synthesis of RNA to the regulation of G-coupled Protein Receptors (GPCR) and cell signaling molecules such as Rho-signaling via Guanine Nucleotide Exchange Factors (GEF).
Solubility
Slightly soluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
27821-45-0 | |
448.12 | |
ZQKVPFKBNNAXCE-HVQNCJTOSA-L | |
131676107 |
C9H12N2Na2O12P2 | |
MFCD00006524,MFCD00084679,MFCD04975448 | |
5'-udp,2na | |
[Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O |
Specifications
27821-45-0 | |
C9H12N2Na2O12P2 | |
100 mg | |
Moisture sensitive | |
Slightly soluble in water. | |
[Na+].[Na+].OC1C(O)[C@@H](O[C@@H]1COP(O)(=O)OP([O-])([O-])=O)N1C=CC(=O)NC1=O | |
448.12 | |
448.13 | |
Powder |
White | |
MFCD00006524,MFCD00084679,MFCD04975448 | |
8817400 | |
5'-udp,2na | |
ZQKVPFKBNNAXCE-HVQNCJTOSA-L | |
disodium 1-[(2R,5R)-3,4-dihydroxy-5-({[hydroxy(phosphonatooxy)phosphoryl]oxy}methyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
131676107 | |
98% | |
Uridine-5'-diphosphate disodium salt |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 248-678-9
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only