Please login to your online account to display your discounted pricing

Urea USP, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Urea
CAS 57-13-6
Purity Grade USP
Molecular Formula NH2CONH2
Formula Weight 60.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK8642500

 
avantor performance
864212
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.06
Quantity 500g
Chemical Name or Material Urea
Purity Grade USP
Molecular Formula NH2CONH2
CAS 57-13-6
Packaging POLYSTORMOR