Please login to your online account to display your discounted pricing

Urea, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$105.61 - $267.23

Chemical Identifiers

Chemical Name or Material Urea
CAS 57-13-6
Purity Grade Ultrapure Bioreagent
Molecular Formula NH2CONH2
Formula Weight 60.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004546

 
avantor performance
411105
Poly Bottle 2.5kg Each for $267.23

02004549

 
avantor performance
411101
Poly Bottle 500g Each for $105.61
Description & Specifications

Specifications

Formula Weight 60.06
Chemical Name or Material Urea
Purity Grade Ultrapure Bioreagent
Molecular Formula NH2CONH2
CAS 57-13-6
Specific Gravity 1L = 1.32kg
Quantity 2.5kg
Packaging Poly Bottle

    For Protein Solubilization and Denaturation