missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Urea, ultrapure, 99%

Manufacturer:  Alfa Aesar™ J6576922

 View more versions of this product

Catalog No. AAJ6576922



Description

Description

Applications
Urea is a raw fabric for the manufacture of organic resins and is used as protein denaturant sealant and in agriculture, dyes, fuels, functional fluids, intermediates, paint additives and coating additives. Urea is used as a nitrogen-release fertilizer, on-corroding alternative to rock salt for road de-icing, browning agent in factory-produced pretzels, cloud seeding agent, a deep eutectic solvent and type of ionic liquid.

Solubility
Soluble in waterSoluble in water, alcohol, methanol, glycerol, benzene and concentrated hydrochloric acid. Practically insoluble in chloroform and ether.

Notes
Stable at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

Urea
Ammonia-like
132°C to 135°C
9.2 to 9.5
Ultrapure
NH2CONH2
635724
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
1176
60.06
99%
1.335
White
100g
57-13-6
CH4N2O
MFCD00008022
14,9867
Soluble in waterSoluble in water,alcohol,methanol,glycerol,benzene and concentrated hydrochloric acid; Practically insoluble in chloroform and ether.
C(=O)(N)N
60.056
CHEBI:48376
Crystalline

Chemical Identifiers

57-13-6
60.056
XSQUKJJJFZCRTK-UHFFFAOYSA-N
1176
urea
CH4N2O
MFCD00008022
alphadrate, carbamide, carbamimidic acid, carbonyldiamide, carbonyldiamine, isourea, pseudourea, ureaphil, ureophil, urevert
CHEBI:48376
C(=O)(N)N
Safety and Handling

Safety and Handling

EINECSNumber : 200-315-5

RTECSNumber : YR6250000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents