Learn More
Urea hydrogen peroxide adduct, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1394014
Description
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUrea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.
Solubility
Soluble in water.
Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
Specifications
Urea hydrogen peroxide adduct | |
∽90°C (decomposition) | |
CH6N2O3 | |
MFCD00013119 | |
UN1511 | |
Very Hygroscopic | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
hydrogen peroxide;urea | |
31294 | |
94.07 |
124-43-6 | |
Odorless | |
H2NCONH2·H2O2 | |
25 g | |
3680414 | |
14,1782 | |
Soluble in water. | |
C(=O)(N)N.OO | |
94.07 | |
CHEBI:75178 | |
97% |
Chemical Identifiers
124-43-6 | |
94.07 | |
AQLJVWUFPCUVLO-UHFFFAOYSA-N | |
31294 | |
hydrogen peroxide;urea |
CH6N2O3 | |
MFCD00013119 | |
urea hydrogen peroxide, carbamide peroxide, percarbamide, urea peroxide, urea dioxide, hydrogen peroxide urea, urea hydroperoxide, hydroperit, hydroperite, percarbamid | |
CHEBI:75178 | |
C(=O)(N)N.OO |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: UREA HYDROGEN PEROXIDE
EINECSNumber : 204-701-4
RTECSNumber : T4860000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only