missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urea (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical

$1,301.40 - $1,301.40

Chemical Identifiers

CAS 57-13-6
Physical Form Solid
Molecular Formula CH4N2O
Linear Formula NH2CONH2
Formula Weight 60.06
MDL Number MFCD00008022
Synonym Carbamide
Chemical Name or Material Urea
Grade BP/EP/JP/USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
U1712 SDS Fisher Scientific Company
U1712
Poly Pail 12kg Each for $1,301.40
Specifications Description & Specifications

Chemical Identifiers

57-13-6
CH4N2O
60.06
Carbamide
BP/EP/JP/USP
Solid
NH2CONH2
MFCD00008022
Urea

Specifications

57-13-6
500ppm max. (BP)
1 EU/mg max. (EP)
0.1% max. (BP)
1 EU/mg max. (USP)
0.002% max. (USP)
0.1% max. (USP)
CH4N2O
NH2CONH2
60.06
132° to 135°C (USP)
Poly Pail
0.10% max. (JP)
0.010% max. (JP)
1.25mmHg at 25°C
Decomposes information: >132°C
White
12kg
99.0 to 100.5% (USP)
Pass Test (BP)
Pass Test (BP)
0.007% max. (JP)
10ppm max. (BP)
Pass Test (BP)
Solid
0.04% max. (USP)
0.04% max. (JP)
1.0% max. (BP)
MFCD00008022
132° to 135°C (USP)
Carbamide
Meets Requirements (BP)
0.1% max. (BP)
1.335g/cm³
7.5 to 9.5
131°C
Urea
BP/EP/JP/USP

Safety and Handling

Emergency Overview
May cause eye, skin, and respiratory tract irritation.
NFPA
Health:1 Flammability:0 Instability:0

Signal Word:CAUTION!