Please login to your online account to display your discounted pricing

Urea (Colorless-to-White Crystals or Crystalline Powder/Mol. Biol.), Fisher BioReagents™


Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP169212

 View more versions of this product

Catalog No. BP169-212

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99%
Formula Weight 60.06
Purity Grade Molecular Biology Grade
Melting Point 131°C
Quantity 2.5kg
Insoluble Matter Pass Test
Physical Form Solid
Color White
pH 7.5 to 9.5
DNase DNase free
Protease Protease free
Vapor Pressure 1.25mmHg at 25°C
Merck Index 15, 10052
Biuret 0.1% max.
Assay Percent Range ≥99 %
Cyanate 5ppm max.
Identification Pass Test
Linear Formula NH2CONH2
Molecular Formula CH4N2O
Heavy Metals (as Pb) 5ppm max.
Iron (Fe) 2ppm max.
Copper (Cu) 1ppm max.
Packaging Poly Bottle
Absorbance 0.05 max. (5M solution) at 280nm
Purity Grade Notes DNase-, RNase- and Protease-Free
CAS 57-13-6
Chemical Name or Material Urea
ChemAlert Storage Symbol Gray
Recommended Storage RT

Specifications

Chemical Name or Material Urea
CAS 57-13-6
Purity Grade Molecular Biology Grade
Purity Grade Notes DNase-, RNase- and Protease-Free
Physical Form Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula CH4N2O
Linear Formula NH2CONH2
Formula Weight 60.06
Accessories
 

RT

SDS & Safety Documents