missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Urea ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 57-13-6
Physical Form Crystalline
Molecular Formula NH2CONH2
Formula Weight 60.06
Chemical Name or Material Urea
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK8648212 Avantor Performance
864806
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57-13-6
NH2CONH2
Urea
Crystalline
60.06
ACS Grade

Specifications

60.06
Crystalline
Poly
Urea
2.5kg
NH2CONH2
57-13-6
ACS Grade