Please login to your online account to display your discounted pricing

Urea ACS AR Crystal, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  864804

 View more versions of this product

Catalog No. MK8648500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Urea
CAS 57-13-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula NH2CONH2
Formula Weight 60.06g/mol

Specifications

Chemical Name or Material Urea
CAS 57-13-6
Purity Grade ACS
Physical Form Crystalline
Molecular Formula NH2CONH2
Formula Weight 60.06g/mol