Please login to your online account to display your discounted pricing

Urea, 99.5%, for analysis, ACROS Organics™


Quantity 5kg

Manufacturer: fisher scientific acros  140750050

 View more versions of this product

Catalog No. AC140750050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fine Crystals
Formula Weight 60.06
Assay ≥99.5% (ex nitrogen)
Quantity 5kg
Color White
Purity Grade For analysis
Melting Point 132° to 135°C
CAS 57-13-6
Insoluble Matter 0.01% max.
Synonym Carbamide
Linear Formula H2NCONH2
Infrared Spectrum Authentic
Chlorine (Cl) 5ppm max.
Ash 0.01% max.
Iron (Fe) 10ppm max.
Heavy Metals (as Pb) 10ppm max.
Chemical Name or Material Urea
Sulfate 10ppm max.
Assay Percent Range 99.5%
Molecular Formula CH4N2O

Specifications

Chemical Name or Material Urea
CAS 57-13-6
Purity Grade For analysis
Assay ≥99.5% (ex nitrogen)
Assay Percent Range 99.5%
Molecular Formula CH4N2O
Linear Formula H2NCONH2
Formula Weight 60.06
Synonym Carbamide