Please login to your online account to display your discounted pricing

URB 597, Tocris™

Potent and selective FAAH inhibitor

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material URB 597
Assay Percent Range >99%
Molecular Formula C20H22N2O3
Formula Weight 338.4
Synonym Cyclohexylcarbamic acid 3'-(Aminocarbonyl)-[1, 1'-biphenyl]-3-yl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

461250

 
r & d systems
RDS461250
50mg Each for $762.58

461210

 
r & d systems
RDS461210
10mg Each for $181.19
Description & Specifications

Specifications

Formula Weight 338.4
Quantity 50mg
Chemical Name or Material URB 597
Assay Percent Range >99%
Molecular Formula C20H22N2O3
CAS 546141-08-6
Synonym Cyclohexylcarbamic acid 3'-(Aminocarbonyl)-[1, 1'-biphenyl]-3-yl ester
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C