Please login to your online account to display your discounted pricing

Urazole 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  140740050

 View more versions of this product

Catalog No. AC140740050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 101.06
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Merck Index 15, 10051
Beilstein 26, 192
TSCA TSCA
EINECS Number 221-776-9
Melting Point 243° to 248°C
Infrared Spectrum Authentic
Synonym 1, 2, 4-Triazolidine-3
Name Note 97%
Assay Percent Range 97%
CAS 3232-84-6
Chemical Name or Material Urazole
Molecular Formula C2 H3 N3 O2
MDL Number MFCD00005225

Specifications

Chemical Name or Material Urazole
Name Note 97%
CAS 3232-84-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2 H3 N3 O2
Formula Weight 101.06
MDL Number MFCD00005225
Synonym 1, 2, 4-Triazolidine-3