Please login to your online account to display your discounted pricing

Urazole 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Urazole
Name Note 97%
CAS 3232-84-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2 H3 N3 O2
Formula Weight 101.06
MDL Number MFCD00005225
Synonym 1, 2, 4-Triazolidine-3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC140740050

  SDS
thermo fisher scientific
140740050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 101.06
Physical Form Crystalline Powder
Quantity 5g
Color White to off-white
Beilstein 26, 192
Merck Index 15, 10051
EINECS Number 221-776-9
TSCA TSCA
Melting Point 243° to 248°C
Chemical Name or Material Urazole
Name Note 97%
Assay Percent Range 97%
Infrared Spectrum Authentic
Molecular Formula C2 H3 N3 O2
MDL Number MFCD00005225
CAS 3232-84-6
Synonym 1, 2, 4-Triazolidine-3