missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Uranium AA Standard, 1mL = 1mg U (1,000ppm U)UO3 in 3% HNO3, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7697-37-2,1344-58-7
CAS Min % 93.0,4.76,0.11
CAS Max % 97.0,4.95,0.12
Physical Form Liquid
Chemical Name or Material Uranium AA Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AU1KN-500 Ricca Chemical Company
RIAU1KN500
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AU1KN-100 Ricca Chemical Company
RIAU1KN100
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7697-37-2,1344-58-7
97.0,4.95,0.12
Uranium AA Standard
93.0,4.76,0.11
Liquid

Specifications

7732-18-5,7697-37-2,1344-58-7
97.0,4.95,0.12
1mL = 1mg U (1, 000ppm U)UO3 in 3% HNO3
Clear yellow
Uranium AA Standard
93.0,4.76,0.11
Liquid
Poly natural
500mL

Safety and Handling

ShelfLife : 24 months