Please login to your online account to display your discounted pricing

Alfa Aesar™ Uracil-6-carboxaldehyde monohydrate, 98%

$105.78 - $363.14

Chemical Identifiers

Chemical Name or Material Uracil-6-carboxaldehyde monohydrate
CAS 36327-91-0
Molecular Formula C5H4N2O3·H2O
Formula Weight 158.11 (140.10 Anhydrous)
MDL Number MFCD03002359
Synonym 6-Formyluracil monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5046903

  SDS
alfa aesar™
H5046903
1g Each for $363.14

AAH50469MD

  SDS
alfa aesar™
H50469MD
250mg Each for $105.78
Description & Specifications

Specifications

Formula Weight 158.11 (140.10 Anhydrous)
Synonym 6-Formyluracil monohydrate
Molecular Formula C5H4N2O3·H2O
CAS 36327-91-0
Quantity 1g
MDL Number MFCD03002359
Assay 98%
Chemical Name or Material Uracil-6-carboxaldehyde monohydrate

Applications
Uracil-6-carboxaldehyde monohydrate is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.