Please login to your online account to display your discounted pricing

Tocris™ UPF 1069

PARP-2 inhibitor

$244.81 - $997.23

Specifications

Product Type UPF 1069
Inhibitors UPF 1069
Molecular Weight 279.29
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

373610R

 
tocris™
RDS373610
10mg Each for $244.81

373650R

 
tocris™
RDS373650
50mg Each for $997.23
Description & Specifications

Specifications

Product Type UPF 1069
Inhibitors UPF 1069
Molecular Weight 279.29
Purity >99

Selective poly(ADP-ribose) polymerase (PARP) 2 inhibitor (IC50 values are 0.3 and 8.0μM for PARP-2 and PARP-1 respectively).

  • Chemical Name: 5-(2-Oxo-2-phenylethoxy)-3,4-dihydroisoquinolin-1(2H)-one
  • Formula: C17H13NO3
  • CAS Number: 1048371-03-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99