Upchurch Scientific™ Valves
$259.56 - $274.39
Specifications
Diameter (English) Inner 0.04 in.
Pressure 500psi
Width (English) 0.66 in.
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Length (English) Swept Volume Price Quantity  
Catalog Number Mfr. No. Length (English) Swept Volume Price Quantity  
05-704-18 Upchurch Scientific™
UPV100L*
1.80 in. 6.2μL Each for $261.31
05-704-20 Upchurch Scientific™
UPV100T
1.80 in. 9.7μL Each for $259.56
05-704-24 Upchurch Scientific™
UPV101L
2.18 in. 6.2μL Each for $274.39
Specifications
Description
Warranty
One-year parts and labor
Description & Specifications
Specifications
0.04 in.
500psi
0.66 in.