Idex Threaded Port Adapters DFS Item
Threaded port adapters help bring products with different thread configurations together
Manufacturer:  Idex UPP621
 View more versions of this product
Catalog No. 05-700-150
Specifications
Specifications
1/4-28 Flat Bottom to M6 Flat Bottom
Tefzel (ETFE)
Not Applicable
1000 psi
Documents