Idex Threaded Port Adapters DFS Item
Threaded port adapters help bring products with different thread configurations together
$14.07 - $70.52
Specifications
Pressure 1000 psi
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (English) Inner Material Thread Size Swept Volume Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Material Thread Size Swept Volume Price Quantity  
05-700-156
View Documents
Idex
UPP627
0.02 in. PEEK 10-32 to 1/4-28 0.30 µL Each for $61.74
05-700-155
View Documents
Idex
UPP626
0.02 in. PEEK 10-32 Coned to M6 Flat Bottom 0.30 µL Each for $70.52
05-700-150
View Documents
Idex
UPP621
Not Applicable Tefzel (ETFE) 1/4-28 Flat Bottom to M6 Flat Bottom Each for $14.07
05-700-325
View Documents
Idex
UPP654
0.03 in. PEEK 10-32 to 1/4-28 6.7 µL Each for $44.87
05-700-342
View Documents
Idex
UPP670
0.03 in. Kel-F 1/4-28 Flat Bottom to M6 Flat Bottom 2.55 µL Each for $60.26
Specifications
Specifications
1000 psi