missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Idex Health & Science™ Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID DFS Item

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Manufacturer:  Idex Health & Science™ UPU147

 View more versions of this product

Catalog No. 05-701-138



Description

Description

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Specifications

9 in.
316 Stainless Steel
Undesignated
1/16 in.
5,800 psi (400 bar)
20 cm
Tubing
0.046 in.
1.15 mm
Documents