Idex Stainless Steel Ferrules DFS Item
Valco/VICI Compatible
Manufacturer:  Idex U401X
 View more versions of this product
Catalog No. 05-700-230
Description
Description
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar).
Specifications
Specifications
ROHS
0.38 cm
20000 psi
Ferrule
0.062 in. O.D. Tubing
0.15 in.
Stainless Steel
10 to 32 coned
10/Pk.
Documents