Idex Stainless Steel Ferrules DFS Item
Valco/VICI Compatible
Supplier:  Idex UPU321X
 View more versions of this product
Catalog No. 05-700-224
Description
Description
Stainless steel fittings withstand pressures up to 20,000 psi (1380 bar).
Specifications
Specifications
0.06 in. OD Tubing
10 to 32 coned
ROHS
10/Pk.
20,000 psi
Stainless Steel
Valco Male Hex Nut