Idex Quick Connect Adapter DFS Item
Convenience of quick connect and disconnect of a luer connection
Manufacturer:  Idex UPP659
 View more versions of this product
Catalog No. 05-700-335
Description
Description
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications
Specifications
Tan
PEEK
0.05 in.
Female Luer to Female 10-32 Adapter
Female Luer Adapter
Documents