Upchurch Scientific™ Quick Connect Adapter DFS Item
Manufacturer:  Upchurch Scientific™ JUP676
 View more versions of this product
Catalog No. 05-700-365
Specifications
Specifications
Male Luer Adapter
0.05 in.
Natural
Pair to Female 10-32
ETFE
40°C
Documents