Upchurch Scientific™ Precolumn Inline Filters
$131.85 - $293.61
Specifications
Products ${productFamilyLength}
Catalog Number Mfr. No. Housing Material Mass Pore Size Price Quantity  
Catalog Number Mfr. No. Housing Material Mass Pore Size Price Quantity  
05-707-101 Upchurch Scientific™
UPA411
PEEK Swept Volume: 103μL 10μm Each for $293.61
05-707-30 Upchurch Scientific™
UPA315*
Stainless Steel Swept Volume: 0.66μL 2μm Each for $131.85
05-707-31 Upchurch Scientific™
UPA316*
Stainless Steel Swept Volume: 0.66μL 0.5μm Each for $148.57
05-707-32 Upchurch Scientific™
UPA318
Stainless Steel Swept Volume: 0.23μL 0.5μm Each for $169.57
29-008-999 Upchurch Scientific™
UPA356
PEEK Swept Volume: 1.3μL 0.5μm Each for $144.81
Specifications
Description
Upchurch Scientific™ Precolumn Inline Filters
Description & Specifications
Specifications
0.0