Upchurch Scientific™ Precolumn Inline Filters DFS Item
Manufacturer:  Upchurch Scientific™ A356*
 View more versions of this product
Catalog No. 05-701-400
Description
Description
Upchurch Scientific™ Precolumn Inline Filters
Specifications
Specifications
0.5μm
PEEK
Swept Volume: 1.3μL
Precolumn Inline Filter
Documents