Idex Plastic Tubing Cutter DFS Item
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
Supplier:  Idex UPA328
 View more versions of this product
Catalog No. 05-701-85
Description
Description
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
  • Built-in guide holes for both 1/16 and 1/8 in. OD tubing
  • Replacement blades included and sold separately
Flat, 90°, burr-free cut
Offers durability, reliability and ease
  • Able to cut through polymers such as PEEK™, FEP, PFA and Tefzel™
  • Specifications
    Specifications
    Replacement Blade
    Steel
    Upchurch™ standard polymer tubing cutter
    Silver