Upchurch Scientific™ PEEK™ Sample Loops
Offer consistent volumes due to tight tolerances of extrusion processes
$113.28 - $119.31
Specifications
For Use With (Application) Analytical instrumentation, in-vitro diagnostics, biotechnology, laboratory
Material PEEK Polymer
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Volume (Metric) Sample Price Quantity  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Volume (Metric) Sample Price Quantity  
05-701-421 Upchurch Scientific™
1815
0.03 in. 0.07cm 500μL Each for $113.28
05-701-422 Upchurch Scientific™
1820
0.03 in. 0.07cm 1mL Each for $119.31
Specifications
Description
PEEK polymer sample loops are biocompatible and serve as an alternative to stainless steel used in life-science applications.
  • Resistant to chemicals and solvents
  • Compatible with Rheodyne™ and VICI™ (Valco) analytical injection valves
  • Flexibe loops for fitting any injection valve
  • Available in volumes ranging from 2μL to 20mL
  • Volumes stated at ±10%, with exact calibration services available
Note: 1/16 in. O.D. sample loops are shipped with F-287 SealTight Fittings, which are pressure rated to 9,000 psi (620 bar)†
Each calibrated sample loop is documented and supplied with calibration certificate.
Analytical instrumentation, in vitro Diagnostics, Biotechnology, Laboratory
† This pressure rating reflects the performance of the fitting, not the tubing or port. Note that the pressure rating of the fitting is well beyond the maximum recommended operating pressure rating of PEEK polymer tubing.
Description & Specifications
Specifications
Analytical instrumentation, in-vitro diagnostics, biotechnology, laboratory
PEEK Polymer