missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Idex One-Piece, High-Pressure Fingertight Fittings: PEEK DFS Item

Fingertight coned fittings can withstand up to 9000 psi.

$156.00 - $156.00

Specifications

Type Fingertight One-Piece Fitting
For Use With (Equipment) 1/16 in. OD Tubing
Material PEEK
View More Specs

Products 2
Catalog Number Mfr. No. Color Length (English) Pressure Rating Price Quantity  
Catalog Number Mfr. No. Color Length (English) Pressure Rating Price Quantity  
05-700-3 DFS Item
View Documents Promotion Details
Idex
UPF130X*
Natural 1.03 in. 6000 psi
Pack of 10 Each for $156.00
 
05-700-1 DFS Item
View Documents Promotion Details
Idex
UPF120X
Natural 0.82 in. 5000 psi
Pack of 10 Each for $156.00
 
Description

Description

Fingertight coned fittings can withstand up to 9000 psi.

Fingertight fittings offer the convenience of making a tubing connection without the need for wrenches or specialized tools when tightening the fitting into place

  • Pressures up to 7000psi (483 bar)
  • Temperatures up to 125°C
  • Fingertights for 1/16 in. OD and smaller tubing
  • Replaceable Ferrules for two-piece versions

Specifications

Specifications

Fingertight One-Piece Fitting
1/16 in. OD Tubing
PEEK
SDS
Product Certifications