Idex Male-Threaded Nuts DFS Item
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Manufacturer:  Idex UPP331*
 View more versions of this product
Catalog No. 29-008-1376
Description
Description
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Specifications
Specifications
Natural
PEEK
0.25 in.
0.365 in.
1/8 in. O.D. Tubing
Nut
0.87 in.
Documents