Upchurch Scientific™ LuerTight™ Fittings: Tefzel DFS Item
Manufacturer:  Upchurch Scientific™ UPP629
 View more versions of this product
Catalog No. 05-700-158
Specifications
Specifications
Luer Adapter
0.04 in.
Natural
To Female 10-32
ETFE
80°C
Documents