missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Upchurch Scientific™ Low-Pressure Unions: Tefzel DFS Item

Manufacturer:  Upchurch Scientific™ UPP710

 View more versions of this product

Catalog No. 05-703-14



Specifications

Specifications

Union
3.17mm
Natural
w/tubing up to 3.17mm O.D.
ETFE
80°C
Documents